Phenolrot ACS
Lieferant: Thermo Scientific
Achtung
Synonyme: Phenolsulfonphthalein, 4,4-(3H-2,1-Benzoxathiol-3- yliden)bisphenol-1,1-dioxid)
16294.06 16294.14 16294.22
16294.06EA 35.7 EUR
16294.06 16294.14 16294.22
Phenolrot ACS
Phenolrot
CAS No.: 143-74-8 Molecular Formula: C19H14O5S Formula Weight: 354.38 Physical Form: Crystalline Odour: Odourless Melting Point: 300° Beilstein Reference: 326470 Merck Reference: 14,7247 MDL No.: MFCD00003552
pH indicator
Formel: C₁₉H₁₄O₅S
Molecular Weight: 354,38 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003552
CAS-Nummer: 143-74-8
EINECS: 205-609-7
Merck Index: 14,7247