4-Heptylbiphenyl
Lieferant: Thermo Scientific
Synonyme:
4-n-Heptylbiphenyl
BTB09570.1
BTB09570.10
BTB09570.50
MAYBBTB09570.1EA
0
EUR
MAYBBTB09570.1
MAYBBTB09570.10
MAYBBTB09570.50
4-Heptylbiphenyl
4-Heptylbiphenyl
Formel:
C₁₉H₂₄ Molecular Weight: 252.4 g/mol Flammpunkt: >110 °C |
CAS-Nummer:
59662-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...