2,5-Dimethylphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
B23740.03
B23740.06
B23740.14
B23740.03EA
32.4
EUR
B23740.03
B23740.06
B23740.14
2,5-Dimethylphenylboronsäure ≥98%
2,5-Dimethylphenylboronsäure
Purity: 98%
CAS No.: 85199-06-0
Molecular Formula: C8H11BO2
Formula Weight: 149.99
Melting Point: ca 180°
Beilstein Reference: 3241785
Formel:
C₈H₁₁BO₂ Molecular Weight: 149,98 g/mol Schmelzpunkt: ∼ 180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863525 CAS-Nummer: 85199-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...