4,5-Dibrom-o-xylol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1,2-Dibrom-4,5-dimethylbenzol, 4,5-Dibrom-1,2-dimethylbenzol

M-Clarity™ Qualitätsstufe = MQ100

638447-1G 638447-5G
SIAL638447-1GEA 47 EUR
SIAL638447-1G SIAL638447-5G
4,5-Dibrom-o-xylol, Sigma-Aldrich®
4,5-Dibrom-o-xylol
Formel: C₈H₈Br₂
Molecular Weight: 263,96 g/mol
MDL: MFCD00082743
CAS-Nummer: 24932-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR