4-tert-Butyl-1-(brommethyl)benzol, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
1-(Bromomethyl)-4-(tert-butyl)benzene, 4-(tert-Butyl)benzylbromide
M-Clarity™ Qualitätsstufe = MQ200
270768-25G
270768-5G
SIAL270768-25GEA
142
EUR
SIAL270768-25G
SIAL270768-5G
4-tert-Butyl-1-(brommethyl)benzol, Sigma-Aldrich®
4-tert-Butyl-1-(brommethyl)benzol
Formel:
C₁₁H₁₅Br Molecular Weight: 227.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000180 CAS-Nummer: 18880-00-7 EINECS: 242-643-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...