5-Brom-2-iod-m-xylol
Lieferant: FLUOROCHEM
Synonyme:
1-Brom-3,5-dimethyl-4-iodbenzol
018285-5G
018285-25G
FLUO018285-5GEA
45.9
EUR
FLUO018285-5G
FLUO018285-25G
5-Brom-2-iod-m-xylol
5-Brom-2-iod-m-xylol
Formel:
C₈H₈BrI Molecular Weight: 310,96 g/mol |
MDL:
MFCD03094656 CAS-Nummer: 206559-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...