2,4-Dimethylbenzaldehyd ≥90%
Lieferant: Thermo Scientific
Achtung
|
A14655.06
A14655.14
A14655.06EA
31.4
EUR
A14655.06
A14655.14
2,4-Dimethylbenzaldehyd ≥90%
2,4-Dimethylbenzaldehyde
Purity: 90+%
CAS No.: 15764-16-6
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 88°(190°F)
Refractive Index: 1.5490
Odour: Almond-like
Melting Point: -9°
Boiling Point: 215-216°
MDL No.: MFCD00003340
Formel:
C₉H₁₀O Molecular Weight: 134,18 g/mol Siedepunkt: 215…216 °C Schmelzpunkt: –9 °C Dichte: 1,02 Flammpunkt: 88 °C (190 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003340 CAS-Nummer: 15764-16-6 EINECS: 239-856-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...