α,α'-Dichlor-m-xylol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
m-Xyloldichlorid, 1,3-Bis(chlormethyl)benzol, m-Xylylendichlorid
A11548.09
A11548.14
A11548.22
A11548.09EA
44.1
EUR
A11548.09
A11548.14
A11548.22
α,α'-Dichlor-m-xylol ≥97%
α,α'-Dichlor-m-xylol
Purity: 97%
CAS No.: 626-16-4
Molecular Formula: C8H8Cl2
Formula Weight: 175.06
Flash Point: 144°(291°F)
Melting Point: 33-37°
Boiling Point: 250-255°
Beilstein Reference: 2079898
UN No.: 2928
MDL No.: MFCD00000912
Formel:
C₈H₈Cl₂ Molecular Weight: 175,06 g/mol Siedepunkt: 250…255 °C Schmelzpunkt: 33…37 °C Dichte: 1,302 Flammpunkt: 144 °C (291 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000912 CAS-Nummer: 626-16-4 EINECS: 210-932-1 UN: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...