3-Chlor-o-xylol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Chlor-1,2-dimethylbenzol, 1-Chlor-2,3-dimethylbenzol

B21505.14 B21505.22
B21505.14EA 68.6 EUR
B21505.14 B21505.22
3-Chlor-o-xylol ≥97%
3-Chlor-o-xylol

Purity: 97% CAS No.: 608-23-1 Molecular Formula: C8H9Cl Formula Weight: 140.61 MDL No.: MFCD00060689

Formel: C₈H₉Cl
Molecular Weight: 140.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060689
CAS-Nummer: 608-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR