4-Chlor-m-xylol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
B21275.09
B21275.18
B21275.09EA
44.6
EUR
B21275.09
B21275.18
4-Chlor-m-xylol ≥97%
4-Chlor-m-xylol
Purity: 97%
CAS No.: 95-66-9
Molecular Formula: C8H9Cl
Formula Weight: 140.61
MDL No.: MFCD00060644
Formel:
C₈H₉Cl Molecular Weight: 140,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060644 CAS-Nummer: 95-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...