2'-Methylacetophenon 98%
Lieferant: Thermo Scientific
Synonyme:
Methyl-o-tolylketon, o-Methylacetophenon
126160250
126160050
126161000
ACRO126160250EA
69.5
EUR
ACRO126160250
ACRO126160050
ACRO126161000
2'-Methylacetophenon 98%
2'-Methylacetophenon
Formel:
C₉H₁₀O Molecular Weight: 134,18 g/mol Siedepunkt: 214 °C (760 mmHg) Dichte: 1,026 g/cm³ Flammpunkt: 75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008734 CAS-Nummer: 577-16-2 EINECS: 209-408-5 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5290 to 1.5330 (20°C, 589 nm) |
GC | ≥97.5 % |
UV transmittance | ≥99 % (c=0.01M water, 254 nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...