2'-Methylacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl-o-tolylketon, o-Methylacetophenon

A11695.06 A11695.14 A11695.22
A11695.06EA 24.5 EUR
A11695.06 A11695.14 A11695.22
2'-Methylacetophenon ≥98%
2'-Methylacetophenon

Purity: 98% CAS No.: 577-16-2 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 75°(167°F) Refractive Index: 1.5318 Boiling Point: 214° Beilstein Reference: 907005 MDL No.: MFCD00008734

Formel: C₉H₁₀O
Molecular Weight: 134.18 g/mol
Siedepunkt: 214 °C
Dichte: 1.020
Flammpunkt: 75 °C (167 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008734
CAS-Nummer: 577-16-2
EINECS: 209-408-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR