3'-Methylacetophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl-m-tolylketon, m-Methylacetophenon
L00440.06
L00440.14
L00440.06EA
28
EUR
L00440.06
L00440.14
3'-Methylacetophenon ≥97%
3'-Methylacetophenon
Purity: 97%
CAS No.: 585-74-0
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 85°(185°F)
Refractive Index: 1.5310
Melting Point: -9°
Boiling Point: 219°
Beilstein Reference: 956637
MDL No.: MFCD00008742
Formel:
C₉H₁₀O Molecular Weight: 134.18 g/mol Siedepunkt: 219 °C Schmelzpunkt: –9 °C Dichte: 1.010 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008742 CAS-Nummer: 585-74-0 EINECS: 209-561-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...