3,5-Dimethylbenzoylchlorid

Lieferant: FLUOROCHEM
Gefahr

014687-1G 014687-25G
FLUO014687-1GEA 31.1 EUR
FLUO014687-1G FLUO014687-25G
3,5-Dimethylbenzoylchlorid
3,5-Dimethylbenzoylchlorid
Formel: C₉H₉ClO
Molecular Weight: 168.62 g/mol
MDL: MFCD02313742
CAS-Nummer: 6613-44-1
EINECS: 413-010-9
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR