2,3-Dimethylbenzamid

Lieferant: Thermo Scientific

Synonyme: 2,3-Dimethylbenzoesäureamid

TL00288.1 TL00288.10 TL00288.50
MAYBTL00288.1EA 0 EUR
MAYBTL00288.1 MAYBTL00288.10 MAYBTL00288.50
2,3-Dimethylbenzamid
2,3-Dimethylbenzamid
Formel: C₉H₁₁NO
MDL: MFCD00017570
CAS-Nummer: 5580-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR