5-Fluor-m-xylol
Lieferant: FLUOROCHEM
Synonyme:
1,3-Dimethyl-5-fluorbenzol, 1-Fluor-3,5-dimethylbenzol, 5-Fluor-1,3-dimethylbenzol
002576-5G
002576-25G
FLUO002576-5GEA
72.9
EUR
FLUO002576-5G
FLUO002576-25G
5-Fluor-m-xylol
5-Fluor-m-xylol
Formel:
C₈H₉F Molecular Weight: 124.16 g/mol Siedepunkt: 145 |
MDL:
MFCD00052366 CAS-Nummer: 461-97-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...