4'-(Trifluormethyl)propiophenon

Lieferant: FLUOROCHEM

Synonyme: p-(Trifluormethyl)propiophenon

007660-5G 007660-10G
FLUO007660-5GEA 97.2 EUR
FLUO007660-5G FLUO007660-10G
4'-(Trifluormethyl)propiophenon
4'-(Trifluormethyl)propiophenon
Formel: C₁₀H₉F₃O
Molecular Weight: 202.18 g/mol
MDL: MFCD00039231
CAS-Nummer: 711-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR