2,5-Bis(trifluormethyl)benzoylchlorid
Lieferant: CHEMPUR
Synonyme:
2,5-Bis(trifluormethyl)benzoesäurechlorid
FL13761.POR
013761-25G
013761-5G
CHMPFL13761.POREA
0
EUR
CHMPFL13761.POR
FLUO013761-25G
FLUO013761-5G
2,5-Bis(trifluormethyl)benzoylchlorid
2,5-Bis(trifluormethyl)benzoylchlorid
Formel:
C₉H₃ClF₆O Molecular Weight: 276,57 g/mol |
MDL:
MFCD00074997 CAS-Nummer: 393-82-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...