2,6-Bis(trifluormethyl)benzoylchlorid

Lieferant: FLUOROCHEM

006019-1G 006019-5G
FLUO006019-1GEA 83.7 EUR
FLUO006019-1G FLUO006019-5G
2,6-Bis(trifluormethyl)benzoylchlorid
2,6-Bis(trifluormethyl)benzoylchlorid
Formel: C₉H₃ClF₆O
Molecular Weight: 276,57 g/mol
MDL: MFCD00061228
CAS-Nummer: 53130-44-2
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR