3,5-Bis(trifluormethyl)benzoylchlorid
Lieferant: Thermo Scientific
Synonyme:
3,5-Bis(trifluormethyl)benzoesäurechlorid
DP01899.1
DP01899.10
DP01899.50
MAYBDP01899.1EA
0
EUR
MAYBDP01899.1
MAYBDP01899.10
MAYBDP01899.50
3,5-Bis(trifluormethyl)benzoylchlorid
3,5-Bis(trifluormethyl)benzoylchlorid
Formel:
C₉H₃ClF₆O Siedepunkt: 80…81 °C (24 mmHg) Dichte: 1,53 g/cm³ (19 °C) |
MDL:
MFCD00000387 CAS-Nummer: 785-56-8 EINECS: 212-322-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...