2,6-Bis(trifluormethyl)benzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B20486.03 B20486.06
B20486.03EA 36.5 EUR
B20486.03 B20486.06
2,6-Bis(trifluormethyl)benzoesäure ≥98%
2,6-Bis(trifluormethyl)benzoesäure

Purity: 98% CAS No.: 24821-22-5 Molecular Formula: C9H4F6O2 Formula Weight: 258.12 Odour: Odourless Melting Point: 136-140° Beilstein Reference: 2057459 MDL No.: MFCD00000376

Formel: C₉H₄F₆O₂
Molecular Weight: 258.12 g/mol
Siedepunkt: 260
Schmelzpunkt: 136…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000376
CAS-Nummer: 24821-22-5
EINECS: 246-479-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR