2,6-Bis(trifluormethyl)benzoesäure

Lieferant: FLUOROCHEM

CFF822. 001948-5G 001948-10G 001948-1G
CHMPCFF822.EA 0 EUR
CHMPCFF822. FLUO001948-5G FLUO001948-10G FLUO001948-1G
2,6-Bis(trifluormethyl)benzoesäure
2,6-Bis(trifluormethyl)benzoesäure
Formel: C₉H₄F₆O₂
Molecular Weight: 258.12 g/mol
Siedepunkt: 260
Schmelzpunkt: 138…139 °C
MDL: MFCD00000376
CAS-Nummer: 24821-22-5
EINECS: 246-479-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR