3,5-Bis(trifluormethyl)phenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1,3-Bis(trifluormethyl)-5-isothiocyanatobenzol

002558-5G 002558-25G
FLUO002558-5GEA 42.9 EUR
FLUO002558-5G FLUO002558-25G
3,5-Bis(trifluormethyl)phenylisothiocyanat
3,5-Bis(trifluormethyl)phenylisothiocyanat
Formel: C₉H₃F₆NS
Molecular Weight: 271.19 g/mol
Siedepunkt: 94…95 °C (19 mmHg)
Dichte: 1.48 g/cm³ (20 °C)
MDL: MFCD00040838
CAS-Nummer: 23165-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR