α,α,α,α',α',α'-Hexafluor-2,4-xylidin

Lieferant: FLUOROCHEM

Synonyme: 2,4-Bis(trifluormethyl)anilin

007828-5G 007828-1G
FLUO007828-5GEA 368 EUR
FLUO007828-5G FLUO007828-1G
α,α,α,α',α',α'-Hexafluor-2,4-xylidin
α,α,α,α',α',α'-Hexafluor-2,4-xylidin
Formel: C₈H₅F₆N
Molecular Weight: 229.12 g/mol
Siedepunkt: 50 °C (0.4 mmHg)
MDL: MFCD00236209
CAS-Nummer: 367-71-5
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR