4-Fluor-3-(trifluormethyl)benzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
(4-Fluoro-3-(trifluoromethyl)phenyl)methanol
006415-1G
006415-5G
FLUO006415-1GEA
56.7
EUR
FLUO006415-1G
FLUO006415-5G
4-Fluor-3-(trifluormethyl)benzylalkohol
4-Fluor-3-(trifluormethyl)benzylalkohol
Formel:
C₈H₆F₄O Molecular Weight: 194.13 g/mol |
MDL:
MFCD00061171 CAS-Nummer: 67515-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...