3,5-Dimethylphenylisocyanat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3,5-Xylylisocyanat

L11698.03 L11698.06
L11698.03EA 27.5 EUR
L11698.03 L11698.06
3,5-Dimethylphenylisocyanat ≥97%
3,5-Dimethylphenylisocyanat

Purity: 97% CAS No.: 54132-75-1 Molecular Formula: C9H9NO Formula Weight: 147.18 Storage Temperature: 2°C to 8°C Flash Point: 83°(181°F) Refractive Index: 1.5280 Boiling Point: 205° Beilstein Reference: 774958 UN No.: 2206 MDL No.: MFCD00013868

Formel: C₉H₉NO
Siedepunkt: 205 °C
Dichte: 1,045
Flammpunkt: 83 °C (181 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00013868
CAS-Nummer: 54132-75-1
EINECS: 258-987-0
UN: 2206
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR