2,6-Diethylphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1,3-Diethyl-2-isothiocyanatobenzene
008865-5G
008865-25G
FLUO008865-5GEA
112
EUR
FLUO008865-5G
FLUO008865-25G
2,6-Diethylphenylisothiocyanat
2,6-Diethylphenylisothiocyanat
Formel:
C₁₁H₁₃NS Molecular Weight: 191.3 g/mol Siedepunkt: 82 °C (0.3 mmHg) |
MDL:
MFCD00041069 CAS-Nummer: 25343-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...