4-tert-Butylbenzylamin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
p-tert-Butylbenzylamine
M-Clarity™ Qualitätsstufe = MQ100
631280-25G
631280-5G
SIAL631280-25GEA
204
EUR
SIAL631280-25G
SIAL631280-5G
4-tert-Butylbenzylamin, Sigma-Aldrich®
4-tert-Butylbenzylamin
Formel:
C₁₁H₁₇N Molecular Weight: 163,26 g/mol |
MDL:
MFCD00040754 CAS-Nummer: 39895-55-1 EINECS: 254-681-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...