6-Nitro-2,4-xylidin

Lieferant: CHEMPUR

Synonyme: 2,4-Dimethyl-6-nitroanilin

BTB10518.250 BTB10518.1
CHMPBTB10518.250EA 0 EUR
CHMPBTB10518.250 MAYBBTB10518.1
6-Nitro-2,4-xylidin
6-Nitro-2,4-xylidin
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
MDL: MFCD00007739
CAS-Nummer: 1635-84-3
EINECS: 216-662-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR