4-tert-Butylbenzylalkohol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B21260.06
B21260.14
B21260.22
B21260.06EA
38.5
EUR
B21260.06
B21260.14
B21260.22
4-tert-Butylbenzylalkohol ≥98%
4-tert-Butylbenzylalkohol
Purity: 98%
CAS No.: 877-65-6
Molecular Formula: C11H16O
Formula Weight: 164.25
Flash Point: 128°(262°F)
Refractive Index: 1.5170
Boiling Point: 139-140°/20mm
Beilstein Reference: 1860665
MDL No.: MFCD00004662
Formel:
C₁₁H₁₆O Siedepunkt: 139…140 °C (20 mmHg) Dichte: 0,928 Flammpunkt: 128 °C (262 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004662 CAS-Nummer: 877-65-6 EINECS: 212-894-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...