2-Methylbenzylalkohol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(Hydroxymethyl)toluole, (2-Methylphenyl)methanol
A12696.09
A12696.18
A12696.30
A12696.09EA
38.1
EUR
A12696.09
A12696.18
A12696.30
2-Methylbenzylalkohol ≥98%
2-Methylbenzylalkohol
Purity: 98%
CAS No.: 89-95-2
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 104°(219°F)
Refractive Index: 1.5408
Melting Point: 33-37°
Boiling Point: 219°
Beilstein Reference: 1929783
MDL No.: MFCD00004622
Formel:
C₈H₁₀O Molecular Weight: 122,17 g/mol Siedepunkt: 219 °C Schmelzpunkt: 33…37 °C Dichte: 1,08 g/cm³ (20 °C) Flammpunkt: 104 °C (219 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004622 CAS-Nummer: 89-95-2 EINECS: 201-954-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...