Thymol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-Isopropyl-m-kresol, 2-Isopropyl-5-methylphenol, 4-Isopropyl-3-hydroxytoluol
A14563.22
A14563.30
A14563.36
A14563.22EA
30.8
EUR
A14563.22
A14563.30
A14563.36
Thymol ≥98%
Thymol
Purity: 98+%
CAS No.: 89-83-8
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 102°(215°F)
pH: 5-7
Melting Point: 49-52°
Boiling Point: 231-233°
Beilstein Reference: 1907135
Merck Reference: 14,9399
UN No.: 2430
MDL No.: MFCD00002309
Formel:
2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH Molecular Weight: 150,22 g/mol Siedepunkt: 231…233 °C Schmelzpunkt: 49…52 °C Dichte: 0,970 Flammpunkt: 102 °C (215 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002309 CAS-Nummer: 89-83-8 EINECS: 201-944-8 UN: 2430 ADR: 8,III Merck Index: 14,9399 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...