4,4'-Bis(dimethylamino)benzophenon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Michlers Keton, 4,4'-Di(dimethylamino)benzophenon

A11889.22 A11889.36
A11889.22EA 38.8 EUR
A11889.22 A11889.36
4,4'-Bis(dimethylamino)benzophenon ≥98%
4,4'-Bis(dimethylamino)benzophenon

Purity: 98% CAS No.: 90-94-8 Molecular Formula: C17H20N2O Formula Weight: 268.36 Flash Point: 220°(428°F) Melting Point: 174-176° Boiling Point: 360° Beilstein Reference: 790733 Merck Reference: 14,6177 MDL No.: MFCD00008312

Formel: C₁₇H₂₀N₂O
Molecular Weight: 268,36 g/mol
Siedepunkt: >360 °C
Schmelzpunkt: 174…176 °C
Flammpunkt: 220 °C (428 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008312
CAS-Nummer: 90-94-8
EINECS: 202-027-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,6177

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR