2-Brom-1,3,5-triisopropylbenzol 96%
Synonyme:
1-Brom-2,4,6-triisopropylbenzol
A15692.14
A15692.22
A15692.36
A15692.14EA
88.3
EUR
A15692.14
A15692.22
A15692.36
2-Brom-1,3,5-triisopropylbenzol 96%
2-Brom-1,3,5-triisopropylbenzol
Purity: 96%
CAS No.: 21524-34-5
Molecular Formula: C15H23Br
Formula Weight: 283.25
Refractive Index: 1.5225
Boiling Point: 125-126°/4mm
MDL No.: MFCD00051547
Formel:
C₁₅H₂₃Br Molecular Weight: 283,25 g/mol Siedepunkt: 125…126 °C (4 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051547 CAS-Nummer: 21524-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...