3',4'-Dimethylacetophenon ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-(3,4-Dimethylphenyl)ethan-1-on
A12187.06
A12187.14
A12187.22
A12187.06EA
26.7
EUR
A12187.06
A12187.14
A12187.22
3',4'-Dimethylacetophenon ≥98%
3',4'-Dimethylacetophenon
Purity: 98%
CAS No.: 3637-01-2
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 102°(215°F)
Refractive Index: 1.5380
Boiling Point: 242-243°
Beilstein Reference: 907053
MDL No.: MFCD00008743
Formel:
C₁₀H₁₂O Molecular Weight: 148.2 g/mol Siedepunkt: 242…243 °C Schmelzpunkt: –3 °C Dichte: 1.002 Flammpunkt: 102 °C (215 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008743 CAS-Nummer: 3637-01-2 EINECS: 222-859-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...