2',6'-Dimethylacetophenon

Lieferant: abcr

Synonyme: 1-(2,6-Dimethylphenyl)ethan-1-on

C-2142-76.0010 C-2142-76.0050
ABCRC-2142-76.0010EA 0 EUR
ABCRC-2142-76.0010 ABCRC-2142-76.0050
2',6'-Dimethylacetophenon
2',6'-Dimethylacetophenon
Formel: C₁₀H₁₂O
Molecular Weight: 148.2 g/mol
MDL: MFCD01861394
CAS-Nummer: 2142-76-9
EINECS: 218-400-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR