2',5'-Dimethylacetophenon ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-(2,5-Dimethylphenyl)ethan-1-on
A11432.06
A11432.14
A11432.06EA
34.1
EUR
A11432.06
A11432.14
2',5'-Dimethylacetophenon ≥98%
2',5'-Dimethylacetophenon
Purity: 98+%
CAS No.: 2142-73-6
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 95°(203°F)
Refractive Index: 1.5290
Odour: Characteristic
Boiling Point: 230-231°
Beilstein Reference: 2042139
MDL No.: MFCD00014988
Formel:
C₁₀H₁₂O Molecular Weight: 148.2 g/mol Siedepunkt: 231 Schmelzpunkt: –18 °C Dichte: 1.0154 g/cm³ (0 °C) Flammpunkt: 95 °C (203 °F) |
MDL:
MFCD00014988 CAS-Nummer: 2142-73-6 EINECS: 218-399-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...