2-Iodmesitylen ≥98%

Lieferant: Thermo Scientific

Synonyme: 1-Iod-2,4,6-trimethylbenzol, 2-Iod-1,3,5-trimethylbenzol, 2,4,6-Trimethyliodbenzol

A15360.06 A15360.14
A15360.06EA 50.9 EUR
A15360.06 A15360.14
2-Iodmesitylen ≥98%
2-Iodmesitylen

Purity: 98% CAS No.: 4028-63-1 Molecular Formula: C9H11I Formula Weight: 246.09 Melting Point: 28-30° Beilstein Reference: 1860064

Formel: C₉H₁₁I
Molecular Weight: 246,09 g/mol
Schmelzpunkt: 28…30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013707
CAS-Nummer: 4028-63-1
EINECS: 223-709-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR