2,4-Dimethylbenzylamin 98%

Lieferant: Thermo Scientific
Gefahr

365530010 365530050
ACRO365530010EA 48.2 EUR
ACRO365530010 ACRO365530050
2,4-Dimethylbenzylamin 98%
2,4-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
Siedepunkt: 218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025575
CAS-Nummer: 94-98-4
EINECS: 202-380-5
UN: 2735
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5360 to 1.5390 (20°C, 589 nm)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR