2,5-Dimethylbenzylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (2,5-Dimethylphenyl)methanamine

350860010 350860050
ACRO350860010EA 13.2 EUR
ACRO350860010 ACRO350860050
2,5-Dimethylbenzylamin 98%
2,5-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135.21 g/mol
MDL: MFCD00039780
CAS-Nummer: 93-48-1
EINECS: 202-250-8
UN: 2735

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5380 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR