3,5-Dimethylbenzylamin

Lieferant: FLUOROCHEM

008879-1G 008879-5G
FLUO008879-1GEA 84.9 EUR
FLUO008879-1G FLUO008879-5G
3,5-Dimethylbenzylamin
3,5-Dimethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135.21 g/mol
MDL: MFCD00040753
CAS-Nummer: 78710-55-1
EINECS: 278-974-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR