3,4-Dimethylbenzylalkohol ≥97%
Lieferant: Thermo Scientific
Synonyme:
4-(Hydroxymethyl)-o-xylene, (3,4-Dimethylphenyl)methanol
B24926.06
B24926.14
B24926.06EA
102
EUR
B24926.06
B24926.14
3,4-Dimethylbenzylalkohol ≥97%
3,4-Dimethylbenzylalkohol
Purity: 97%
CAS No.: 6966-10-5
Molecular Formula: C9H12O
Formula Weight: 136.19
Melting Point: 62-65°
Boiling Point: 218-221°
MDL No.: MFCD00004647
Formel:
C₉H₁₂O Molecular Weight: 136,19 g/mol Siedepunkt: 218…221 °C Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004647 CAS-Nummer: 6966-10-5 EINECS: 230-175-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...