2,4-Dimethylbenzylalkohol ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
(2,4-Dimethylphenyl)methanol, 4-(Hydroxymethyl)-m-xylene
L02899.03
L02899.06
L02899.03EA
34.5
EUR
L02899.03
L02899.06
2,4-Dimethylbenzylalkohol ≥97%
2,4-Dimethylbenzylalkohol
Purity: 97%
CAS No.: 16308-92-2
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point:
Formel:
C₉H₁₂O Molecular Weight: 136.19 g/mol Siedepunkt: 120…121 °C (13 mmHg) Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004624 CAS-Nummer: 16308-92-2 EINECS: 240-393-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...