2,4,6-Trimethylthiophenol

Lieferant: Thermo Scientific

Synonyme: 2,4,6-Trimethylphenylmercaptan

F02-12350 BTB12350DA BTB12350EA BTB12350.50 BTB12350-0001
MAYBF02-12350EA 0 EUR
MAYBF02-12350 MAYBBTB12350DA MAYBBTB12350EA MAYBBTB12350.50 MAYBBTB12350-0001
2,4,6-Trimethylthiophenol
2,4,6-Trimethylthiophenol
Formel: C₉H₁₂S
MDL: MFCD00218475
CAS-Nummer: 1541-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR