2-Brombiphenyl (PBB Nr. 1) ≥98%
Lieferant: Thermo Scientific
Synonyme:
1-Brom-2-phenylbenzol, PBB Nr. 1
B21840.03
B21840.06
B21840.14
B21840.03EA
30.9
EUR
B21840.03
B21840.06
B21840.14
2-Brombiphenyl (PBB Nr. 1) ≥98%
2-Brombiphenyl (PBB Nr. 1)
Purity: 98%
CAS No.: 2052-07-5
Molecular Formula: C12H9Br
Formula Weight: 233.12
Flash Point:
Formel:
C₁₂H₉Br Molecular Weight: 233,11 g/mol Siedepunkt: 298 °C Schmelzpunkt: 0…2 °C Dichte: 1,376 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000065 CAS-Nummer: 2052-07-5 EINECS: 218-141-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...