3-Brombiphenyl (PBB Nr. 2) ≥99%

Lieferant: Thermo Scientific

Synonyme: 1-Brom-3-phenylbenzol

B21726.03 B21726.06 B21726.14
B21726.03EA 74.4 EUR
B21726.03 B21726.06 B21726.14
3-Brombiphenyl (PBB Nr. 2) ≥99%
3-Brombiphenyl (PBB Nr. 2)

Purity: 99% CAS No.: 2113-57-7 Molecular Formula: C12H9Br Formula Weight: 233.12 Flash Point: 110°(230°F) Refractive Index: 1.6410 Boiling Point: 301° Beilstein Reference: 2043191 MDL No.: MFCD00000082

Formel: C₁₂H₉Br
Molecular Weight: 233,11 g/mol
Siedepunkt: 301 °C
Schmelzpunkt: 9 °C
Dichte: 1,397
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000082
CAS-Nummer: 2113-57-7
EINECS: 218-304-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR