4,4'-Dibrombiphenyl (PBB Nr. 15) 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
PBB Nr. 15
212081000
212085000
212080250
ACRO212081000EA
67
EUR
ACRO212081000
ACRO212085000
ACRO212080250
4,4'-Dibrombiphenyl (PBB Nr. 15) 99%
4,4'-Dibrombiphenyl (PBB Nr. 15)
Formel:
C₁₂H₈Br₂ Molecular Weight: 312 g/mol Siedepunkt: 357 Schmelzpunkt: 165…169 °C Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000101 CAS-Nummer: 92-86-4 EINECS: 202-198-6 UN: 3152 ADR: 9 II |
Spezifikation Testergebnisse
Appearance | White to yellow to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 165 to 169 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...