4-Brombiphenyl (PBB Nr. 3)
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1-Brom-4-phenylbenzol, PBB Nr. 3
024187-100G
024187-25G
FLUO024187-100GEA
195
EUR
FLUO024187-100G
FLUO024187-25G
4-Brombiphenyl (PBB Nr. 3)
4-Brombiphenyl (PBB Nr. 3)
Formel:
C₁₂H₉Br Molecular Weight: 233.11 g/mol Siedepunkt: 310 Schmelzpunkt: 90…91 °C Dichte: 1.615 g/cm³ (20 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000100 CAS-Nummer: 92-66-0 EINECS: 202-176-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...