Benzyl-4-hydroxybenzoat ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzylparaben, 4-Hydroxybenzoesäurebenzylester
A11735.09
A11735.18
A11735.36
A11735.09EA
36.8
EUR
A11735.09
A11735.18
A11735.36
Benzyl-4-hydroxybenzoat ≥99%
Benzyl-4-hydroxybenzoat
Purity: 99%
CAS No.: 94-18-8
Molecular Formula: C14H12O3
Formula Weight: 228.25
Melting Point: 110-114°
Beilstein Reference: 2115995
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228,25 g/mol Schmelzpunkt: 110…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016471 CAS-Nummer: 94-18-8 EINECS: 202-311-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...