2-Fluorbiphenyl ≥98%

Lieferant: Thermo Scientific

Synonyme: PFB Nr. 1, 1-Fluor-2-phenylbenzol

A12227.06 A12227.09 A12227.14
A12227.06EA 71.9 EUR
A12227.06 A12227.09 A12227.14
2-Fluorbiphenyl ≥98%
2-Fluorbiphenyl

Purity: 98% CAS No.: 321-60-8 Molecular Formula: C12H9F Formula Weight: 172.20 Melting Point: 72-76° Boiling Point: 248° MDL No.: MFCD00000317

Formel: C₁₂H₉F
Molecular Weight: 172,2 g/mol
Siedepunkt: 248 °C
Schmelzpunkt: 72…76 °C
Dichte: 1,245
Lagertemperatur: Raumtemperatur
MDL: MFCD00000317
CAS-Nummer: 321-60-8
EINECS: 206-290-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR